Thermo Scientific Chemicals

4-Methoxybenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific Chemicals

Catalog number: H54078.AE
100 mL, Each
Thermo Scientific Chemicals

4-Methoxybenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific Chemicals

Catalog number: H54078.AE
100 mL, Each
Quantity
Catalog number: H54078.AE
also known as H54078-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
38769-92-5
IUPAC Name
chloro[(4-methoxyphenyl)methyl]magnesium
Molecular Formula
C8H9ClMgO
InChI Key
XHDYUCUVUZSISM-UHFFFAOYSA-M
SMILES
COC1=CC=C(C[Mg]Cl)C=C1
Form
Liquid
Concentration
0.2-0.3M

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text