2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%, Thermo Scientific Chemicals
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H54088.MD
also known as H54088-MD
Price (USD)/ Each
62.40
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
62.40
Add to cart
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog numberH54088.MD
Price (USD)/ Each
62.40
-
Add to cart
Specifications
Chemical Name or Material2-(Trifluoromethyl)pyrimidine-5-carboxylic acid
CAS306960-77-0
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)pyrimidine-5-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only