Thermo Scientific Chemicals

1,3-Dioxane, 98%, Thermo Scientific Chemicals

Catalog number: H54107.14
25 g, Each
Thermo Scientific Chemicals

1,3-Dioxane, 98%, Thermo Scientific Chemicals

Catalog number: H54107.14
25 g, Each
Quantity
Catalog number: H54107.14
also known as H54107-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
505-22-6
IUPAC Name
1,3-dioxane
Molecular Formula
C4H8O2
InChI Key
VDFVNEFVBPFDSB-UHFFFAOYSA-N
SMILES
C1COCOC1
Form
Liquid
Purity
≥97.5%
Appearance (Color)
Colorless to almost colorless

Description

1,3-Dioxane has been employed as solvent in the synthesis of 8- and 9-membered dioxazocines and dioxazonines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dioxane has been employed as solvent in the synthesis of 8- and 9-membered dioxazocines and dioxazonines.

Solubility
Fully miscible in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text