Thermo Scientific Chemicals

(Boc-aminooxy)acetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: H54109.03
1 g, Each
Thermo Scientific Chemicals

(Boc-aminooxy)acetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: H54109.03
1 g, Each
Quantity
Catalog number: H54109.03
also known as H54109-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
42989-85-5
IUPAC Name
2-({[(tert-butoxy)carbonyl]amino}oxy)acetic acid
Molecular Formula
C7H13NO5
InChI Key
QBXODCKYUZNZCY-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NOCC(O)=O
Assay (HPLC)
>98.0%

Description

(Boc-aminooxy)acetic acid was employed to introduce a hydroxylamine moiety into peptides; reaction with an aldehyde to an oxime. It was used in the preparation of Boc-aminooxy tetra(ethylene glycol).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Boc-aminooxy)acetic acid was employed to introduce a hydroxylamine moiety into peptides; reaction with an aldehyde to an oxime. It was used in the preparation of Boc-aminooxy tetra(ethylene glycol).

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text