Thermo Scientific Chemicals

5-Fluoroindoline, 97%, Thermo Scientific Chemicals

Catalog number: H54158.03
1 g, Each
Thermo Scientific Chemicals

5-Fluoroindoline, 97%, Thermo Scientific Chemicals

Catalog number: H54158.03
1 g, Each
Quantity
Catalog number: H54158.03
also known as H54158-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2343-22-8
IUPAC Name
5-fluoro-2,3-dihydro-1H-indole
Molecular Formula
C8H8FN
InChI Key
NXQRMQIYCWFDGP-UHFFFAOYSA-N
SMILES
FC1=CC2=C(NCC2)C=C1
Form
Oil
Assay (unspecified)
>96.0%
Appearance (Color)
Colorless to light green or yellow

Description

5-Fluoroindoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoroindoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text