Specifications
Chemical Name or Material2,4-Dichlorobenzylmagnesium chloride
CAS129752-86-9
Health Hazard 1H225-H302-H314-H335
Health Hazard 2GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
View more