Chemical Identifiers
CAS125700-73-4
IUPAC Name[(dimethylamino)({3,5-dioxo-4-azatricyclo[5.2.1.0²,⁶]dec-8-en-4-yl}oxy)methylidene]dimethylazanium; tetrafluoroboranuide
Molecular FormulaC14H20BF4N3O3
InChI KeyDMTWWFCCBWWXSA-UHFFFAOYNA-N
SMILESF[B-](F)(F)F.CN(C)C(ON1C(=O)C2C3CC(C=C3)C2C1=O)=[N+](C)C
View more