Benzofurazan-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Benzofurazan-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzofurazan-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H54275.MD
also known as H54275-MD
Price (USD)/ Each
67.80
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
67.80
Add to cart
Benzofurazan-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog numberH54275.MD
Price (USD)/ Each
67.80
-
Add to cart
Specifications
Chemical Name or MaterialBenzofurazan-4-sulfonyl chloride
CAS114322-14-4
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only