Thermo Scientific Chemicals

Methyl pyrrole-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H54340.03
1 g, Each
Thermo Scientific Chemicals

Methyl pyrrole-3-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: H54340.03
1 g, Each
Quantity
Catalog number: H54340.03
also known as H54340-03
Price (USD)
Quantity
-
Have Questions?

Description

Methyl pyrrole-3-carboxylate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl pyrrole-3-carboxylate is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text