(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 99+%, Thermo Scientific Chemicals
(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H54378.03
also known as H54378-03
Price (USD)/ Each
80.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
80.50
Add to cart
(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 99+%, Thermo Scientific Chemicals
Catalog numberH54378.03
Price (USD)/ Each
80.50
-
Add to cart
Chemical Identifiers
CAS156311-83-0
IUPAC Namehexafluoro-λ⁵-phosphanuide; tris(pyrrolidin-1-yl)({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})phosphanium
Molecular FormulaC17H27F6N7OP2
InChI KeyCBZAHNDHLWAZQC-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.C1CCN(C1)[P+](ON1N=NC2=CC=CN=C12)(N1CCCC1)N1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>99.0%
(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate is used as reagent for synthesis of cyclic RGC pentapeptides for functionalization through click chemistry, fluorescent glucose bioprobes and ReactIR™ flow cell. It is also used as reagent for disulfide bond engineering, heteroaryl ether synthesis and optimization of conformational constraint on non-phosphorylated cyclic peptide antagonists of the Grb2-SH2 domain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate is used as reagent for synthesis of cyclic RGC pentapeptides for functionalization through click chemistry, fluorescent glucose bioprobes and ReactIR™ flow cell. It is also used as reagent for disulfide bond engineering, heteroaryl ether synthesis and optimization of conformational constraint on non-phosphorylated cyclic peptide antagonists of the Grb2-SH2 domain.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Hyang Yeon Lee; Jae Jeong Lee; Jongmin Park; Seung Bum Park. Development of fluorescent glucose bioprobes and their application on real-time and quantitative monitoring of glucose uptake in living cells. Chemistry: A European Journal. 2011, 17, (1),143-150
  2. Markus Muttenthaler; Asa Andersson; Aline D de Araujo; Zoltan Dekan; Richard J.Lewis; Paul F.Alewood. Modulating oxytocin activity and plasma stability by disulfide bond engineering. Journal of medicinal and pharmaceutical chemistry. 2010, 53, (24),8585-8596