5-Bromo-8-methoxyquinoline, 96%, Thermo Scientific Chemicals
5-Bromo-8-methoxyquinoline, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromo-8-methoxyquinoline, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H54386.MD
also known as H54386-MD
Price (USD)/ Each
55.70
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
55.70
Add to cart
5-Bromo-8-methoxyquinoline, 96%, Thermo Scientific Chemicals
Catalog numberH54386.MD
Price (USD)/ Each
55.70
-
Add to cart
Specifications
Chemical Name or Material5-Bromo-8-methoxyquinoline
CAS10522-47-1
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
5-Bromo-8-methoxyquinoline is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-8-methoxyquinoline is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
RUO – Research Use Only

General References:

  1. Tammy Wang, David R.Magnin; Lawrence G.Hamann. Palladium-catalyzed microwave-assisted amination of 1-bromonaphthalenes and 5- and 8-bromoquinolines. Org. Lett. 2003, 5,(6), 897-900.
  2. Yi Cui; Suning Wang. Diboron and triboron compounds based on linear and star-shaped conjugated ligands with 8-hydroxyquinolate functionality: Impact of intermolecular interaction and boron coordination on luminescence. J. Org. Chem. 2006, 71,(17), 6485-6496.