Thermo Scientific Chemicals

Methyl hydrogen terephthalate, 99+%, Thermo Scientific Chemicals

Catalog number: H54391.14
25 g, Each
Thermo Scientific Chemicals

Methyl hydrogen terephthalate, 99+%, Thermo Scientific Chemicals

Catalog number: H54391.14
25 g, Each
Quantity
Catalog number: H54391.14
also known as H54391-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1679-64-7
IUPAC Name
4-(methoxycarbonyl)benzoic acid
Molecular Formula
C9H8O4
InChI Key
REIDAMBAPLIATC-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(C=C1)C(O)=O
Assay (HPLC)
>99.0%

Description

Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.

Solubility
Slightly soluble in water. Also soluble in methanol, benzene and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text