N-Boc-1H-pyrazole-1-carboxamidine, 98+%, Thermo Scientific Chemicals
N-Boc-1H-pyrazole-1-carboxamidine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-1H-pyrazole-1-carboxamidine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H54394.06
also known as H54394-06
Price (USD)/ Each
85.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
85.10
Add to cart
N-Boc-1H-pyrazole-1-carboxamidine, 98+%, Thermo Scientific Chemicals
Catalog numberH54394.06
Price (USD)/ Each
85.10
-
Add to cart
Chemical Identifiers
CAS152120-61-1
IUPAC Nametert-butyl N-[amino(1H-pyrazol-1-yl)methylidene]carbamate
Molecular FormulaC9H14N4O2
InChI KeyIGSFMHYSWZUENI-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N=C(N)N1C=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only