N-(tert-Butoxycarbonyloxy)succinimide, 98+%, Thermo Scientific Chemicals
N-(tert-Butoxycarbonyloxy)succinimide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-(tert-Butoxycarbonyloxy)succinimide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number H54493.14
also known as H54493-14
Price (USD)/ Each
391.65
Online exclusive
435.00 
Save 43.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
391.65
Online exclusive
435.00 
Save 43.35 (10%)
Add to cart
N-(tert-Butoxycarbonyloxy)succinimide, 98+%, Thermo Scientific Chemicals
Catalog numberH54493.14
Price (USD)/ Each
391.65
Online exclusive
435.00 
Save 43.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13139-12-3
IUPAC Nametert-butyl 2,5-dioxopyrrolidin-1-yl carbonate
Molecular FormulaC9H13NO5
InChI KeyVTGFSVGZCYYHLO-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)ON1C(=O)CCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)98.0% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only