2-Bromo-4-methylpyrimidine, 97%, Thermo Scientific Chemicals
2-Bromo-4-methylpyrimidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-4-methylpyrimidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Catalog number H54503.06
also known as H54503-06
Price (USD)/ Each
258.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
258.00
Add to cart
2-Bromo-4-methylpyrimidine, 97%, Thermo Scientific Chemicals
Catalog numberH54503.06
Price (USD)/ Each
258.00
-
Add to cart
Specifications
Chemical Name or Material2-Bromo-4-methylpyrimidine
CAS130645-48-6
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
2-Bromo-4-methylpyrimidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-4-methylpyrimidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Veli-Matti Mukkala; Christian Sund; Marek Kwiatkowski; Paavo Pasanen; Marita Hogberg; Jouko Kankare; Harri Takal. New heteroaromatic complexing agents and luminescence of their europium(III) and terbium(III) chelates. Helvetica Chimica Acta. 1992, 75,(5), 1621-1632.