Thermo Scientific Chemicals

2-(4-Nitrophenyl)ethanol, 98%, Thermo Scientific Chemicals

Catalog number: H54504.14
25 g, Each
Thermo Scientific Chemicals

2-(4-Nitrophenyl)ethanol, 98%, Thermo Scientific Chemicals

Catalog number: H54504.14
25 g, Each
Quantity
Catalog number: H54504.14
also known as H54504-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-27-6
IUPAC Name
2-(4-nitrophenyl)ethan-1-ol
Molecular Formula
C8H9NO3
InChI Key
IKMXRUOZUUKSON-UHFFFAOYSA-N
SMILES
OCCC1=CC=C(C=C1)[N+]([O-])=O
Assay from Supplier's CofA
≥97.5%
Form
Powder and/or Lumps
Appearance (Color)
Yellow to yellow-orange to brown

Description

It is a phosphate protecting reagent and O6 of deoxyguanosine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a phosphate protecting reagent and O6 of deoxyguanosine.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents. Keep at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text