It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Soluble in water at 122.5°Cat760mmHg.
Notes
Hygroscopic. Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only
Michael Green.; Judd Berman. Preparation of pentafluorophenyl esters of FMOC protected amino acids with pentafluorophenyl trifluoroacetate. Tetrahedron Lett. 1990, 31 (41),5851-5852.
Tirayut Vilaivana.; Chaturong Suparpprom.; Pongchai Harnyuttanakorn.; Gordon Lowe. Synthesis and properties of novel pyrrolidinyl PNA carrying β-amino acid spacers. Tetrahedron Lett. 2001, 42 (32),5533-5536.