Thermo Scientific Chemicals

Methyl picolinate, 99%, Thermo Scientific Chemicals

Catalog number: H54530.14
25 g, Each
Thermo Scientific Chemicals

Methyl picolinate, 99%, Thermo Scientific Chemicals

Catalog number: H54530.14
25 g, Each
Quantity
Catalog number: H54530.14
also known as H54530-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2459-07-6
IUPAC Name
methyl pyridine-2-carboxylate
Molecular Formula
C7H7NO2
InChI Key
NMMIHXMBOZYNET-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=N1
Form
Low melting solid
Assay (HPLC)
>98.5%
Appearance (Color)
Colorless to yellow

Description

Methyl picolinate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl picolinate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text