It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Sparingly Soluble (0.062 g/L) (25°C), Calc.
Notes
Incompatible with oxidizing agents. Keep at room temperature.
RUO – Research Use Only
Jelena Božilović, Jan W Bats.; Joachim W Engels. Synthesis and structure of fluoroindole nucleosides. Can. J. Chem.. 2007, 85 (4),283-292.
Kevin McClay.; Baojie Wan.; Yuehong Wang.; Sanghyun Cho.; Jerry Yu.; Bernard Santarsiero.; Shahila Mehboob.; Michael Johnson.; Scott Franzblau. A novel combinatorial biocatalytic approach for producing antibacterial compounds effective against Mycobacterium tuberculosis (TB). Appl. Microbiol. Biotechnol.. 2013, 97 (16),7151-7163.