Thermo Scientific Chemicals

N,N'-Di-Boc-1H-pyrazole-1-carboxamidine, 98+%, Thermo Scientific Chemicals

Catalog number: H54565.06
5 g, Each
Thermo Scientific Chemicals

N,N'-Di-Boc-1H-pyrazole-1-carboxamidine, 98+%, Thermo Scientific Chemicals

Catalog number: H54565.06
5 g, Each
Quantity
Catalog number: H54565.06
also known as H54565-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
152120-54-2
IUPAC Name
tert-butyl N-({[(tert-butoxy)carbonyl]imino}(1H-pyrazol-1-yl)methyl)carbamate
Molecular Formula
C14H22N4O4
InChI Key
QFNFDHNZVTWZED-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)NC(=NC(=O)OC(C)(C)C)N1C=CC=N1
Assay (HPLC)
98.0% min

Description

N,N'-Di-Boc-1H-pyrazole-1-carboxamidine is used in the stereoselective synthesis of the bicyclic guanidine alkaloid (+)-monanchorin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Di-Boc-1H-pyrazole-1-carboxamidine is used in the stereoselective synthesis of the bicyclic guanidine alkaloid (+)-monanchorin.

Solubility
Slightly soluble in water. Soluble in methanol, and chloroform.

Notes
Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text