Thermo Scientific Chemicals

Benzyloxyacetic acid, 95%, Thermo Scientific Chemicals

Catalog number: H54581.06
5 g, Each
Thermo Scientific Chemicals

Benzyloxyacetic acid, 95%, Thermo Scientific Chemicals

Catalog number: H54581.06
5 g, Each
Quantity
Catalog number: H54581.06
also known as H54581-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
30379-55-6
IUPAC Name
2-(benzyloxy)acetic acid
Molecular Formula
C9H10O3
InChI Key
GRZHHTYDZVRPIC-UHFFFAOYSA-N
SMILES
OC(=O)COCC1=CC=CC=C1
Appearance (Color)
Colorless to yellow
Form
Liquid
Assay (GC)
>94.0%

Description

It is a compound for proteomics research use. It may be used for following synthesis mixed benzyloxyacetic pivalic anhydride, chiral glycolates, 1,2:4,5-di-O-isopropylidene-β-D-fructopyranos-3-yl benzyloxyacetate, via esterification, tetra aqua(1,10-phenanthroline-[κ]2N,N?)magnesium(II) bis[(2,4-dichlorophenyl)acetate], as ligand in the preparation of diaquabis(benzyloxyacetato)copper(II) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a compound for proteomics research use. It may be used for following synthesis mixed benzyloxyacetic pivalic anhydride, chiral glycolates, 1,2:4,5-di-O-isopropylidene-β-D-fructopyranos-3-yl benzyloxyacetate, via esterific
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text