Thermo Scientific Chemicals

Indole-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H54638.03
1 g, Each
Thermo Scientific Chemicals

Indole-2-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H54638.03
1 g, Each
Quantity
Catalog number: H54638.03
also known as H54638-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
19005-93-7
Appearance (Color)
Dark yellow
Form
Crystalline solid
Assay (unspecified)
>96.0%

Description

Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.

Solubility
Soluble in Methanol.

Notes
Air sensitive. Strong oxidizers and strong reducing agents are incompatible. Store at room temperature.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text