1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%, Thermo Scientific Chemicals
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H54647.MD
also known as H54647-MD
Price (USD)/ Each
40.60
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
40.60
Add to cart
1-Chloroacetyl-2-(trifluoroacetyl)hydrazine, 97%, Thermo Scientific Chemicals
Catalog numberH54647.MD
Price (USD)/ Each
40.60
-
Add to cart
Chemical Identifiers
CAS762240-99-3
IUPAC Name2-chloro-N'-(2,2,2-trifluoroacetyl)acetohydrazide
Molecular FormulaC4H4ClF3N2O2
InChI KeyDYKIVKLXFDNBMY-UHFFFAOYSA-N
SMILESFC(F)(F)C(=O)NNC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormSolid
Purity≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only