Thermo Scientific Chemicals
1,3-Benzodioxole-5-sulfonyl chloride, 95%, Thermo Scientific Chemicals
Catalog number: H54755.06
5 g, Each
Thermo Scientific Chemicals
1,3-Benzodioxole-5-sulfonyl chloride, 95%, Thermo Scientific Chemicals
Catalog number: H54755.06
5 g, Each
Quantity
Catalog number: H54755.06
also known as H54755-06
Price (USD)
Quantity
-
Have Questions?
Description
1,3-Benzodioxole-5-sulfonyl chloride is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,3-Benzodioxole-5-sulfonyl chloride is used as an organic chemical synthesis intermediate.
Notes
Moisture sensitive. Incompatible with oxidizing agents.