Coumarin-6-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Coumarin-6-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Coumarin-6-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H54767.03
also known as H54767-03
Price (USD)/ Each
48.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
48.20
Add to cart
Coumarin-6-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Catalog numberH54767.03
Price (USD)/ Each
48.20
-
Add to cart
Specifications
Chemical Name or MaterialCoumarin-6-sulfonyl chloride
CAS10543-42-7
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Coumarin-6-sulfonyl chloride is used as a derivatizing agent for amines and phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coumarin-6-sulfonyl chloride is used as a derivatizing agent for amines and phenols.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Fakhr Eldin O.Suliman; Suad S. Al-Kindi; Salma M.Z. Al-Kindy; Haider A.J. Al-Lawati. Analysis of phenols in water by high-performance liquid chromatography using coumarin-6-sulfonyl chloride as a fluorogenic precolumn label. Journal of Chromatography A. 2006, 1101,(1-2), 179-184.
  2. S.M Al-Kindy; F.E.Suliman; A.A Al-Hamadi. Fluorescence enhancement of coumarin-6-sulfonyl chloride amino acid derivatives in cyclodextrin media. Analytical Sciences. 2001, 17,(4), 539-543.