Thermo Scientific Chemicals

Coumarin-6-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H54767.06
5 g, Each
Thermo Scientific Chemicals

Coumarin-6-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H54767.06
5 g, Each
Quantity
Catalog number: H54767.06
also known as H54767-06
Price (USD)
Quantity
-
Have Questions?

Description

Coumarin-6-sulfonyl chloride is used as a derivatizing agent for amines and phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Coumarin-6-sulfonyl chloride is used as a derivatizing agent for amines and phenols.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text