2-Amino-3-methylpyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
2-Amino-3-methylpyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-3-methylpyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H54773.MD
also known as H54773-MD
Price (USD)/ Each
81.90
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
81.90
Add to cart
2-Amino-3-methylpyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Catalog numberH54773.MD
Price (USD)/ Each
81.90
-
Add to cart
Specifications
Chemical Name or Material2-Amino-3-methylpyridine-5-boronic acid pinacol ester
CAS1111637-91-2
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is used as primary and secondary pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Refrigerated (2-4°C). Incompatible with oxidizing agents.
RUO – Research Use Only