Pyrimidine-2-thiocarboxamide, 97%, Thermo Scientific Chemicals
Pyrimidine-2-thiocarboxamide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyrimidine-2-thiocarboxamide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H54783.06
also known as H54783-06
Price (USD)/ Each
472.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
472.00
Add to cart
Pyrimidine-2-thiocarboxamide, 97%, Thermo Scientific Chemicals
Catalog numberH54783.06
Price (USD)/ Each
472.00
-
Add to cart
Specifications
Chemical Name or MaterialPyrimidine-2-thiocarboxamide
CAS4537-73-9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Pyrimidine-2-thiocarboxamide is used as a pharmaceutical intermediate and as a synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrimidine-2-thiocarboxamide is used as a pharmaceutical intermediate and as a synthetic building block.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only