Thermo Scientific Chemicals

1-Methyl-1H-pyrazole-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H54785.MD
250 mg, Each
Thermo Scientific Chemicals

1-Methyl-1H-pyrazole-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H54785.MD
250 mg, Each
Quantity
Catalog number: H54785.MD
also known as H54785-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
288148-34-5
IUPAC Name
1-methyl-1H-pyrazole-4-sulfonyl chloride
Molecular Formula
C4H5ClN2O2S
InChI Key
RDAVKKQKMLINOH-UHFFFAOYSA-N
SMILES
CN1C=C(C=N1)S(Cl)(=O)=O
Assay (GC)
>96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text