Thermo Scientific Chemicals

1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole, 99+%, Thermo Scientific Chemicals

Catalog number: H54792.06
5 g, Each
Thermo Scientific Chemicals

1-(2-Mesitylenesulfonyl)-3-nitro-1H-1,2,4-triazole, 99+%, Thermo Scientific Chemicals

Catalog number: H54792.06
5 g, Each
Quantity
Catalog number: H54792.06
also known as H54792-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
74257-00-4
IUPAC Name
3-nitro-1-(2,4,6-trimethylbenzenesulfonyl)-1H-1,2,4-triazole
Molecular Formula
C11H12N4O4S
InChI Key
SFYDWLYPIXHPML-UHFFFAOYSA-N
SMILES
CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O
Assay (HPLC)
>99.0%

Description

1-(2-Mesitylenesulfonyl)-3-Nitro-1H-1,2,4-Triazole is an efficient coupling reagent in the synthesis of oligonucleotides via phosphotriester approach.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Mesitylenesulfonyl)-3-Nitro-1H-1,2,4-Triazole is an efficient coupling reagent in the synthesis of oligonucleotides via phosphotriester approach.

Solubility
Soluble in Chloroform.

Notes
Store at 4°C. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text