3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt, 97+%, Thermo Scientific Chemicals
3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
25 mg
Catalog number H54807.MC
also known as H54807-MC
Price (USD)/ Each
478.00
-
Add to cart
Quantity:
100 mg
Request bulk or custom format
Price (USD)/ Each
478.00
Add to cart
3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt, 97+%, Thermo Scientific Chemicals
Catalog numberH54807.MC
Price (USD)/ Each
478.00
-
Add to cart
Chemical Identifiers
CAS92921-24-9
IUPAC Namesodium 1-{4-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]cyclohexanecarbonyloxy}-2,5-dioxopyrrolidine-3-sulfonate
Molecular FormulaC16H17N2NaO9S
InChI KeyVUFNRPJNRFOTGK-UHFFFAOYNA-M
SMILES[Na+].[O-]S(=O)(=O)C1CC(=O)N(OC(=O)C2CCC(CN3C(=O)C=CC3=O)CC2)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to white
FormSolid
Purity≥97.0%
3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt is a cross-linking reagent incorporating an extended spacer with amine and sulfhydryl reactivity. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). Second coupling specific for molecules containing free sulfhydryl by thioether linkage buffered at pH 6.8 (6.5-7.0). Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Sulfo-N-succinimidyl 4-(maleimidomethyl)cyclohexane-1-carboxylate sodium salt is a cross-linking reagent incorporating an extended spacer with amine and sulfhydryl reactivity. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). Second coupling specific for molecules containing free sulfhydryl by thioether linkage buffered at pH 6.8 (6.5-7.0). Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates.

Solubility
Soluble in water (2.5 mg/ml) and dimethyl formamide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Wolfgang J. Parak.; Daniele Gerion.; Daniela Zanchet.; Anke S. Woerz.; Teresa Pellegrino.; Christine Micheel.; Shara C. Williams.; Markus Seitz.; Richard E. Bruehl.; Zev Bryant.; Carlos Bustamante.; Carolyn R. Bertozzi.; A. Paul Alivisatos. Conjugation of DNA to Silanized Colloidal Semiconductor Nanocrystalline Quantum Dots. Chem. Mater. 2002, 14 (5), 2113-2119.
  2. Tsuyoshi Tanaka.; Tadashi Matsunaga. Fully Automated Chemiluminescence Immunoassay of Insulin Using Antibody-Protein A-Bacterial Magnetic Particle Complexes. Anal. Chem. 2000, 72 (15), 3518-3522.