Thermo Scientific Chemicals

tert-Pentylmagnesium chloride, 1M in 2-MeTHF, Thermo Scientific Chemicals

Catalog number: H54824.AE
100 mL, Each
Thermo Scientific Chemicals

tert-Pentylmagnesium chloride, 1M in 2-MeTHF, Thermo Scientific Chemicals

Catalog number: H54824.AE
100 mL, Each
Quantity
Catalog number: H54824.AE
also known as H54824-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28276-08-6
IUPAC Name
chloro(2-methylbutan-2-yl)magnesium
Molecular Formula
C5H11ClMg
InChI Key
NWFDYJDCSIHFAB-UHFFFAOYSA-M
SMILES
CCC(C)(C)[Mg]Cl
Assay (GC)
1.0 M (in Methyltetrahydrofuran)

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Store at 4°C. Air and moisture sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text