3,5-Dibromoquinoline, 96%, Thermo Scientific Chemicals
3,5-Dibromoquinoline, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dibromoquinoline, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number H54859.06
also known as H54859-06
Price (USD)/ Each
376.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
376.00
Add to cart
3,5-Dibromoquinoline, 96%, Thermo Scientific Chemicals
Catalog numberH54859.06
Price (USD)/ Each
376.00
-
Add to cart
Chemical Identifiers
CAS101861-59-0
IUPAC Name3,5-dibromoquinoline
Molecular FormulaC9H5Br2N
InChI KeyIBADFXOMCWHDMS-UHFFFAOYSA-N
SMILESBrC1=CC2=C(Br)C=CC=C2N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)White
Assay by NMR>95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only