Thermo Scientific Chemicals

3,5-Dibromoquinoline, 96%, Thermo Scientific Chemicals

Catalog number: H54859.MD
250 mg, Each
Thermo Scientific Chemicals

3,5-Dibromoquinoline, 96%, Thermo Scientific Chemicals

Catalog number: H54859.MD
250 mg, Each
Quantity
Catalog number: H54859.MD
also known as H54859-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
101861-59-0
IUPAC Name
3,5-dibromoquinoline
Molecular Formula
C9H5Br2N
InChI Key
IBADFXOMCWHDMS-UHFFFAOYSA-N
SMILES
BrC1=CC2=C(Br)C=CC=C2N=C1
Form
Solid
Appearance (Color)
White
Assay by NMR
>95.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text