Thermo Scientific Chemicals

4-Fluoro-2-methylphenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific Chemicals

Catalog number: H54865.AE
100 mL, Each
Thermo Scientific Chemicals

4-Fluoro-2-methylphenylmagnesium bromide, 0.50 M in 2-MeTHF, Thermo Scientific Chemicals

Catalog number: H54865.AE
100 mL, Each
Quantity
Catalog number: H54865.AE
also known as H54865-AE
Price (USD)
Quantity
-
Have Questions?

Description

It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in 0.5 M in Tetrahydrofuran.

Notes
Store at 4°C. Air
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text