Chemical Identifiers
CAS126828-35-1
IUPAC Name2-{4-[(2,4-dimethoxyphenyl)({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]phenoxy}acetic acid
Molecular FormulaC32H29NO7
InChI KeyUPMGJEMWPQOACJ-UHFFFAOYNA-N
SMILESCOC1=CC(OC)=C(C=C1)C(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCC(O)=O)C=C1
View more