Thermo Scientific Chemicals

6-Bromo-2-chloroquinoline, 96%, Thermo Scientific Chemicals

Catalog number: H54895.03
1 g, Each
Thermo Scientific Chemicals

6-Bromo-2-chloroquinoline, 96%, Thermo Scientific Chemicals

Catalog number: H54895.03
1 g, Each
Quantity
Catalog number: H54895.03
also known as H54895-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1810-71-5
IUPAC Name
6-bromo-2-chloroquinoline
Molecular Formula
C9H5BrClN
InChI Key
YXRDWUJAJLDABJ-UHFFFAOYSA-N
SMILES
ClC1=NC2=CC=C(Br)C=C2C=C1
Appearance (Color)
White to off-white
Proton NMR
Conforms
Purity
≥95.0%
Form
Solid

Description

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in Chloroform and Methanol

Notes
Store at room temperature with proper ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text