Thermo Scientific Chemicals

Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate, 98%, Thermo Scientific Chemicals

Catalog number: H54910.06
5 g, Each
Thermo Scientific Chemicals

Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate, 98%, Thermo Scientific Chemicals

Catalog number: H54910.06
5 g, Each
Quantity
Catalog number: H54910.06
also known as H54910-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
164298-23-1
IUPAC Name
[(dimethylamino)(fluoro)methylidene]dimethylazanium
Molecular Formula
C5H12FN2
InChI Key
JILUGXGJYPRQPA-UHFFFAOYSA-N
SMILES
CN(C)C(F)=[N+](C)C
Assay (HPLC)
>97.5%

Description

Fluoro-N,N,N',N'-tetramethylformamidinium hexafluorophosphate is used as a reagent for synthesis of cationic antimicrobial β-2,2-amino acid derivatives, philanthotoxin analogues for inhibition of the ionotropic glutamate receptor, small β-peptidomimetics for antimicrobials and hydrazone ligation to assemble multifunctional viral nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate is used as a reagent for synthesis of cationic antimicrobial β-2,2-amino acid derivatives, philanthotoxin analogues for inhibition of the ionotropic glutamate receptor, small β-peptidomimetics for antimicrobials and hydrazone ligation to assemble multifunctional viral nanoparticles.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text