Thermo Scientific Chemicals

Benzylhydrazine dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H54916.06
5 g, Each
Thermo Scientific Chemicals

Benzylhydrazine dihydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: H54916.06
5 g, Each
Quantity
Catalog number: H54916.06
also known as H54916-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20570-96-1
IUPAC Name
dihydrogen benzylhydrazine dichloride
Molecular Formula
C7H12Cl2N2
InChI Key
MSJHOJKVMMEMNX-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NNCC1=CC=CC=C1
Assay (HPLC)
>96.0%

Description

Benzylhydrazine dihydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzylhydrazine dihydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text