It is used in organic synthesis and as an pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in organic synthesis and as an pharmaceutical intermediate.
Solubility
Very soluble in water.
Notes
Store at 4°C. Keep away from oxidizing agents and heat.
RUO – Research Use Only
Quirinus B. Broxterman.; Bernard Kaptein.; Johan Kamphuis.; Hans E. Schoemaker. Synthesis of (optically active) sulfur-containing trifunctional amino acids by radical addition to (optically active) unsaturated amino acids. J. Org. Chem. 1992, 57 (23),6286-6294.
Peng-Fei Xu.; Shuo Li.; Ta-Jung Lu .; Chen-Chang Wu.; Botao Fan.; Georgia Golfis. Asymmetric Synthesis of α,α-Disubstituted α-Amino Acids by Diastereoselective Alkylation of Camphor-Based Tricyclic Iminolactone. J. Org. Chem. 2006, 71 (12),4364-4373.