Thermo Scientific Chemicals

Indole-7-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H54931.06
5 g, Each
Thermo Scientific Chemicals

Indole-7-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H54931.06
5 g, Each
Quantity
Catalog number: H54931.06
also known as H54931-06
Price (USD)
Quantity
-
Have Questions?

Description

Indole-7-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indole-7-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text