Thermo Scientific Chemicals

2,4-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF, Thermo Scientific Chemicals

Catalog number: H54966.AE
100 mL, Each
Thermo Scientific Chemicals

2,4-Difluorobenzylmagnesium bromide, 0.25M in 2-MeTHF, Thermo Scientific Chemicals

Catalog number: H54966.AE
100 mL, Each
Quantity
Catalog number: H54966.AE
also known as H54966-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
546122-71-8
IUPAC Name
bromo[(2,4-difluorophenyl)methyl]magnesium
Molecular Formula
C7H5BrF2Mg
InChI Key
BPKTYCGKUHSSCG-UHFFFAOYSA-M
SMILES
FC1=CC(F)=C(C[Mg]Br)C=C1
Concentration
0.2-0.3M
Comment
Aqueous quench: passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text