Thermo Scientific Chemicals

2-Amino-4-chloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: H55007.03
1 g, Each
Thermo Scientific Chemicals

2-Amino-4-chloropyrimidine, 98%, Thermo Scientific Chemicals

Catalog number: H55007.03
1 g, Each
Quantity
Catalog number: H55007.03
also known as H55007-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3993-78-0
IUPAC Name
4-chloropyrimidin-2-amine
Molecular Formula
C4H4ClN3
InChI Key
DBGFGNCFYUNXLD-UHFFFAOYSA-N
SMILES
NC1=NC=CC(Cl)=N1
Assay (HPLC)
>97.5%

Description

2-Amino-4-chloropyrimidine is a substrate used in a palladium-catalyzed cyanation with zinc(II) cyanide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-chloropyrimidine is a substrate used in a palladium-catalyzed cyanation with zinc(II) cyanide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text