Thermo Scientific Chemicals

2,2-Bis[4-(4-aminophenoxy)phenyl]propane, 98%, Thermo Scientific Chemicals

Catalog number: H55057.22
100 g, Each
Thermo Scientific Chemicals

2,2-Bis[4-(4-aminophenoxy)phenyl]propane, 98%, Thermo Scientific Chemicals

Catalog number: H55057.22
100 g, Each
Quantity
Catalog number: H55057.22
also known as H55057-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13080-86-9
IUPAC Name
4-(4-{2-[4-(4-aminophenoxy)phenyl]propan-2-yl}phenoxy)aniline
Molecular Formula
C27H26N2O2
InChI Key
KMKWGXGSGPYISJ-UHFFFAOYSA-N
SMILES
CC(C)(C1=CC=C(OC2=CC=C(N)C=C2)C=C1)C1=CC=C(OC2=CC=C(N)C=C2)C=C1
Assay (HPLC)
>97.5%

Description

It is applied in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in pharmaceutical industry.

Solubility
Very slightly soluble in water. Soluble in acetone, Dimethyl sulfoxide(DMSO).

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text