Thermo Scientific Chemicals

4-[(Trimethylsilyl)ethynyl]benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H55060.14
25 g, Each
Thermo Scientific Chemicals

4-[(Trimethylsilyl)ethynyl]benzaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H55060.14
25 g, Each
Quantity
Catalog number: H55060.14
also known as H55060-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77123-57-0
IUPAC Name
4-[2-(trimethylsilyl)ethynyl]benzaldehyde
Molecular Formula
C12H14OSi
InChI Key
UZQDUXAJFTWMDT-UHFFFAOYSA-N
SMILES
C[Si](C)(C)C#CC1=CC=C(C=O)C=C1
Assay (GC)
>96.0%

Description

It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, Pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store at 4°C. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text