Thermo Scientific Chemicals

3-Ethyl-4-methyl-3-pyrrolin-2-one, 98%, Thermo Scientific Chemicals

Catalog number: H55065.06
5 g, Each
Thermo Scientific Chemicals

3-Ethyl-4-methyl-3-pyrrolin-2-one, 98%, Thermo Scientific Chemicals

Catalog number: H55065.06
5 g, Each
Quantity
Catalog number: H55065.06
also known as H55065-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
766-36-9
Assay (HPLC)
>97.5%

Description

3-Ethyl-4-methyl-3-pyrrolin-2-one is used as an intermediate for the manufacture of medicines. It is used for the preparation of Glimepiride (G410150), and its intermediates and as antidiabetic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethyl-4-methyl-3-pyrrolin-2-one is used as an intermediate for the manufacture of medicines. It is used for the preparation of Glimepiride (G410150), and its intermediates and as antidiabetic agents.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text