Thermo Scientific Chemicals

2-(Chloromethoxy)ethyltrimethylsilane, tech. 90%, stab. with 0.1% N,N-Diisopropylethylamine, Thermo Scientific Chemicals

Catalog number: H55074.14
25 g, Each
Thermo Scientific Chemicals

2-(Chloromethoxy)ethyltrimethylsilane, tech. 90%, stab. with 0.1% N,N-Diisopropylethylamine, Thermo Scientific Chemicals

Catalog number: H55074.14
25 g, Each
Quantity
Catalog number: H55074.14
also known as H55074-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
76513-69-4
IUPAC Name
[2-(chloromethoxy)ethyl]trimethylsilane
Molecular Formula
C6H15ClOSi
InChI Key
BPXKZEMBEZGUAH-UHFFFAOYSA-N
SMILES
C[Si](C)(C)CCOCCl
Form
Liquid
Comment
Stabilized
Assay (unspecified)
≥88.0%

Description

2-(Chloromethoxy)ethyltrimethylsilane is used in the preparation of SEM [2-(trimethylsilyl)ethoxy]methyl]-ethers. It serves as a phenol protecting group in the synthesis of laterifluorones. Further, it reacts with 1H-imidazole to prepare 1-(2-trimethylsilanyl-ethoxymethyl)-1H-imidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Chloromethoxy)ethyltrimethylsilane is used in the preparation of SEM [2-(trimethylsilyl)ethoxy]methyl]-ethers. It serves as a phenol protecting group in the synthesis of laterifluorones. Further, it reacts with 1H-imidazole to prepare 1-(2-trimethylsilanyl-ethoxymethyl)-1H-imidazole.

Notes
Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text