Thermo Scientific Chemicals

1-Naphthyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: H55091.03
1 g, Each
Thermo Scientific Chemicals

1-Naphthyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

Catalog number: H55091.03
1 g, Each
Quantity
Catalog number: H55091.03
also known as H55091-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99747-74-7
IUPAC Name
naphthalen-1-yl trifluoromethanesulfonate
Molecular Formula
C11H7F3O3S
InChI Key
WQWUQDVFRYMMCY-UHFFFAOYSA-N
SMILES
FC(F)(F)S(=O)(=O)OC1=C2C=CC=CC2=CC=C1
Assay (GC)
>96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text