Thermo Scientific Chemicals

6-Bromoquinoline, 97%, Thermo Scientific Chemicals

Catalog number: H55111.03
1 g, Each
Thermo Scientific Chemicals

6-Bromoquinoline, 97%, Thermo Scientific Chemicals

Catalog number: H55111.03
1 g, Each
Quantity
Catalog number: H55111.03
also known as H55111-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5332-25-2
IUPAC Name
6-bromoquinoline
Molecular Formula
C9H6BrN
InChI Key
IFIHYLCUKYCKRH-UHFFFAOYSA-N
SMILES
BrC1=CC=C2N=CC=CC2=C1
Appearance (Color)
Clear colorless to brown
Form
Liquid
Assay (GC)
≥96.0%

Description

6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromoquinoline is used as fine chemical and pharmaceutical intermediate, used as the coupling reagent.

Solubility
Soluble in acetone, acetonitrile, dichloromethane, ethyl acetate and THF.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text