Thermo Scientific Chemicals

Methyl methanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: H55120.06
5 g, Each
Thermo Scientific Chemicals

Methyl methanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: H55120.06
5 g, Each
Quantity
Catalog number: H55120.06
also known as H55120-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
66-27-3
IUPAC Name
methyl methanesulfonate
Molecular Formula
C2H6O3S
InChI Key
MBABOKRGFJTBAE-UHFFFAOYSA-N
SMILES
COS(C)(=O)=O
Form
Liquid
Appearance (Color)
Colorless to pale yellow
Assay (GC)
>98.5%

Description

Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methanesulfonate is a DNA adduct that adds methyl groups to Dan at 7-guanine, 3-guanine and 3-adenine.

Solubility
Very soluble in water.

Notes
Store at room temperature with proper ventilation. Keep away from oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text